Sodium hydroxide (0.13 - 0.55 mol/l; 0.13 - 0.55 N) in aqueous solution 0.2 N in aqueous solution analytical reagent, VWR Chemicals BDH®

Supplier: VWR International
Warning

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

BDH7220-1 BDH7220-4
BDH7220-1CS 641.97 USD
BDH7220-1 BDH7220-4
Sodium hydroxide (0.13 - 0.55 mol/l; 0.13 - 0.55 N) in aqueous solution 0.2 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Sodium hydroxide

AOAC, APHA, and ASTM. Suitable as titrant.

Formula: NaOH
Boiling Pt: ∼105 °C (1013 hPa)
Density: 1.02 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. traceable to SRM 84 Yes
Normality 0.1995 - 0.2005 N
Shelf-life 18 months


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR