Sodium hydroxide ( 0.13 mol/l; 0.13 N) in aqueous solution 0.1 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Supplier:
VWR International
|
Warning
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
BDH7219-1
BDH7219-4
BDH7219-5
BDH7219-7
BDH7219-1CS
581.39
USD
BDH7219-1
BDH7219-4
BDH7219-5
BDH7219-7
Sodium hydroxide ( 0.13 mol/l; 0.13 N) in aqueous solution 0.1 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Sodium hydroxide
AOAC, APHA, and ASTM. Suitable as titrant.
Formula:
NaOH Density: 1.003 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,III |
Specification Test Results
Appearance/color | Clear liquid |
N.I.S.T. traceable to SRM 84 | Yes |
Normality | 0.0995 - 0.1005 N |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...