Sodium hydroxide 0.02 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

BDH7217-1
BDH7217-1CS 561.02 USD
BDH7217-1
Sodium hydroxide 0.02 N in aqueous solution, VWR Chemicals BDH®
Sodium hydroxide

AOAC, APHA, and ASTM for acidity. Suitable as titrant.

Formula: NaOH
MW: 40 g/mol
Density: 1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. traceable to SRM 84 X
Normality 0.0195 - 0.0205 N
Shelf-life 1 year

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR