Potassium hydroxide 1.0 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

BDH7212-1
BDH7212-1CS 922.79 USD
BDH7212-1
Potassium hydroxide 1.0 N in aqueous solution, VWR Chemicals BDH®
Potassium hydroxide

Made with deionized water. Suitable as a titrant.

Formula: KOH
MW: 56.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,III

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. traceable X
Normality 0.995 - 1.005 N
Shelf-life 2 years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR