Nitric acid solution 1.00 N, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Nitric acid solution

BDH7209-1
BDH7209-1CS 1078.37 USD
BDH7209-1
Nitric acid solution 1.00 N, VWR Chemicals BDH®
Nitric acid

Made with high purity acid and deionized water. Suitable as a titrant.

Formula: HNO₃
MW: 63.01 g/mol
Density: 1.035 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 3264
ADR: 8,III

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. Traceable 1
Normality 0.998 - 1.002 N
Shelf-life 18 months

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR