Iodine solution 0.025 N, VWR Chemicals BDH®

Supplier: VWR International

BDH7206-1
BDH7206-1CS 409.55 USD
BDH7206-1
Iodine solution 0.025 N, VWR Chemicals BDH®
Iodine

Made with iodine crystals, potassium iodide and deionized water.

Formula: I₂
MW: 253.81 g/mol
MDL Number: MFCD00011355
CAS Number: 7553-56-2
UN: 3495
ADR: 8,III

Order Now

Specification Test Results

Normality 0.0200 - 0.0300 N
Appearance/color Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR