Sodium hypochlorite 12.5% in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH7038-4L
BDH7038-4LCS 257.32 USD
BDH7038-4L
Sodium hypochlorite 12.5% in aqueous solution, VWR Chemicals BDH®
Sodium hypochlorite
Formula: ClNaO
Boiling Pt: 111 °C (1013 hPa)
Melting Pt: –30…–20 °C
Density: 1.22…1.25 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00011120
CAS Number: 7681-52-9
EINECS: 231-668-3
UN: 1791
ADR: 8,II

Order Now

Specification Test Results

Assay (W/W) (at preparation) 10.0 - 14.0 %
Appearance Clear yellow/green liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR