1,2,4-Trichlorobenzene ≥99.0%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International

BDH67009.400
BDH67009.400CS 2740.61 USD
BDH67009.400
1,2,4-Trichlorobenzene ≥99.0%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
1,2,4-Trichlorobenzene
Formula: C₆H₃Cl₃
MW: 181.45 g/mol
Boiling Pt: 214.4 °C (1013 hPa)
Melting Pt: 17 °C
Flash Pt: 99 °C
MDL Number: MFCD00000547
CAS Number: 120-82-1
UN: 2321
ADR: 6.1,III

Order Now

Specification Test Results

Assay (by GC-FID) Min 99.0%
Residue after evaporation Max 5 ppm
Water (by KF coulometry) Max 0.01%
Transmittance at 308 nm Min 10 T(%)
Transmittance at 310 nm Min 30 T(%)
Transmittance at 350 nm Min 90 T(%)
Transmittance at 375 nm Min 98 T(%)
Transmittance at 400 nm Min 98 T(%)
Optical absorbance at 308 nm Max 1.00 AU
Optical absorbance at 310 nm Max 0.50 AU
Optical absorbance at 350 nm Max 0.05 AU
Optical absorbance at 375 nm Max 0.01 AU
Optical absorbance at 400 nm Max 0.01 AU

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR