Tetrahydrofuran stabilized, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Oxolane, 1,4-epoxy-Butane, Butylene oxide, Butane alpha,delta-oxide, Tetramethylene oxide, THF, Oxacyclopentane, Cyclotetramethylene oxide, Diethylene oxide

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

BDH67008.400
BDH67008.400CS 2837.25 USD
BDH67008.400
Tetrahydrofuran stabilized, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Tetrahydrofuran
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: –108.5 °C
Flash Pt: –21.5 °C
MDL Number: MFCD00005356
CAS Number: 109-99-9
UN: 2056
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC-FID) Min 99.8%
Color Max 10 APHA
Peroxides/Epoxides (as Hydrogen peroxide at time of packaging) Max 0.015%
Residue after evaporation Max 0.03%
Stabilizer (BHT) 0.02-0.03%
Titrable acid Max 0.001 meq/g
Water (by KF coulometry) Max 0.02%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR