tert-Butyl methyl ether ≥99.8%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: MTBE, 2-Methyl-2-methoxypropane, Ether, tert-butyl methyl, Methyl 1,1-dimethylethyl ether, 2-Methoxy-2-methylpropane, Methyl t-butyl ether, Methyl tert-butyl ether, Methyl-t-Butyl Ether

BDH67005.400
BDH67005.400CS 1584.1 USD
BDH67005.400
tert-Butyl methyl ether ≥99.8%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
tert-Butyl methyl ether
Formula: (CH₃)₃COCH₃
MW: 88.15 g/mol
Boiling Pt: 55.2 °C (1013 hPa)
Melting Pt: –108.6 °C
Flash Pt: –28 °C
MDL Number: MFCD00008812
CAS Number: 1634-04-4
UN: 2398
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC-FID) Min 99.8%
Color Max 10 APHA
Peroxides/Epoxides (as Hydrogen peroxide at time of packaging) Max 1 ppm
Residue after evaporation Max 3 ppm
Water (by KF coulometry) Max 0.5%
Transmittance at 210 nm Min 10 T(%)
Transmittance at 225 nm Min 30 T(%)
Transmittance at 250 nm Min 80 T(%)
Transmittance at 270 nm Min 95 T(%)
Transmittance at 300 nm Min 98 T(%)
Optical absorbance at 210 nm Max 1.00 AU
Optical absorbance at 225 nm Max 0.50 AU
Optical absorbance at 250 nm Max 0.10 AU
Optical absorbance at 270 nm Max 0.02 AU
Optical absorbance at 300 nm Max 0.01 AU

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR