Water, clear, colorless

Supplier: VWR International

BDH6601-4L
BDH6601-4LCS 2000 USD
BDH6601-4L
Water, clear, colorless
Water
Formula: H₂O
Boiling Pt: 100 °C (760 mm Hg)
Melting Pt: 0 °C
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR