EDTA disodium salt dihydrate 99.0-101.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
Synonyms:
Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate
BDH4616-500G
BDH4616-500GCS
915.47
USD
BDH4616-500G
EDTA disodium salt dihydrate 99.0-101.0% ACS, VWR Chemicals BDH®
EDTA disodium salt dihydrate
Powder.
Formula:
C₁₀H₁₄N₂Na₂O₈·2H₂O MW: 372.24 g/mol Melting Pt: 255 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150037 CAS Number: 6381-92-6 EINECS: 205-358-3 Merck Index: 13,03543 |
Specification Test Results
Assay | 99.0 - 101.0 % |
pH (25°C; 5 %) | 4.0 - 6.0 |
Heavy metals (as Pb) | Max. 0.005 % |
Insoluble matter | Max. 0.005 % |
Nitrilotriacetic acid | Max. 0.1 % |
Fe (Iron) | Max. 0.01 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...