Cyclohexanone ≥99.0% ACS, VWR Chemicals BDH®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

BDH4612-4L
BDH4612-4LCS 730.93 USD
BDH4612-4L
Cyclohexanone ≥99.0% ACS, VWR Chemicals BDH®
Cyclohexanone
Formula: C₆H₁₀(=O)
Boiling Pt: 155.6 °C (1013 hPa)
Melting Pt: –31 °C
Density: 0.9478 g/cm³ (20 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00001625
CAS Number: 108-94-1
EINECS: 203-631-1
UN: 1915
ADR: 3,III
REACH: 01-2119453616-35
Merck Index: 13,02755
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8701dac7edb70a452483cd4f1bd716ca29ba9aa&vid=0360daffe7606174162954ba58a6ba2b78e28dfb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.0 %
Color Max. 10 APHA
Residue after evaporation Max. 0.05 %
Water Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR