di-Sodium L(+)-tartrate dihydrate 99.0-101.0% ACS, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Sodium L-(+)-tartrate dihydrate, L(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Sodium tartrate dibasic dihydrate, Disodium L-(+)-Tartrate Dihydrate, (R,R)-(+)-Disodium tartrate dihydrate, L(+)-Tartaric acid, disodium salt, dihydrate, Sodium L-tartrate dibasic dihydrate, L(+)-Sodium tartrate dibasic dihydrate, Sodium Tartrate, Dihydrate, (+)-Disodium L-tartrate dihydrate, (+)-Disodium-L-tartrat dihydrate

BDH4606-500G
BDH4606-500GCS 1159.63 USD
BDH4606-500G
di-Sodium L(+)-tartrate dihydrate 99.0-101.0% ACS, VWR Chemicals BDH®
di-Sodium L(+)-tartrate dihydrate
Formula: NaOOC(CHOH)₂COONa·2H₂O
Melting Pt: 57 °C
Density: 1.818 g/cm³ (20 °C)
MDL Number: MFCD00150035
CAS Number: 6106-24-7
EINECS: 212-773-3
Merck Index: 12,08833
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39a75038f4ff5e6c234267b15540823ae361bed4&vid=89289572c48b0c6415f8f11e916a51f66e129474&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Appearance White crystalline powder
pH (25°C; 5 %) 7.0 - 9.0
Heavy metals (as Pb) Max. 5 ppm
Insoluble matter Max. 0.005 %
Loss on drying (150°C) 15.61 - 15.71 %
Cl (Chloride) Max. 5 ppm
NH4 (Ammonium) Max. 0.003 %
PO4 (Phosphate) Max. 5 ppm
SO4 (Sulphate) Max. 0.005 %
Ca (Calcium) Max. 0.01 %
Fe (Iron) Max. 0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR