Potassium nitrate ≥99.0%, crystallised ACS, VWR Chemicals BDH®

Supplier: VWR International
Warning

BDH4598-500G
BDH4598-500GCS 628.37 USD
BDH4598-500G
Potassium nitrate ≥99.0%, crystallised ACS, VWR Chemicals BDH®
Potassium nitrate
Formula: KNO₃
Melting Pt: 334 °C
Density: 2.109 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011409
CAS Number: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1,III
REACH: 01-2119488224-35
Merck Index: 13,07733

Order Now

Specification Test Results

Assay Min. 99.0 %
pH (25°C; 5 %) 4.5 - 8.5
Heavy metals (as Pb) Max. 5 ppm
Insoluble matter Max. 0.005 %
Cl (Chloride) Max. 0.002 %
IO3 (Iodate) Max. 5 ppm
NO2 (Nitrite) Max. 0.001 %
PO4 (Phosphate) Max. 5 ppm
SO4 (Sulphate) Max. 0.003 %
Ca (Calcium) Max. 0.005 %
Fe (Iron) Max. 3 ppm
Mg (Magnesium) Max. 0.002 %
Na (Sodium) Max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR