Sodium peroxide ≥93.0% ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: di-Sodium peroxide

BDH4584-500G
BDH4584-500GEA 368.27 USD
BDH4584-500G
Sodium peroxide ≥93.0% ACS, VWR Chemicals BDH®
Sodium peroxide
Formula: Na₂O₂
MW: 77.98 g/mol
Melting Pt: 660 °C
Density: 2.805 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003497
CAS Number: 1313-60-6
EINECS: 215-209-4
UN: 1504
ADR: 5.1,I
Merck Index: 13,08728

Order Now

Specification Test Results

Assay Min. 93.0 %
Heavy metals (as Pb) Max. 0.002 %
Cl (Chloride) Max. 0.002 %
PO4 (Phosphate) Max. 5 ppm
SO4 (Sulphate) Max. 0.001 %
Fe (Iron) Max. 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR