4-Methyl-2-pentanone ≥98.5% ACS, VWR Chemicals BDH®

Danger

Synonyms: Methyl iso-Butyl Ketone, Isobutyl Methyl Ketone, Methyl Isobutyl Ketone, MIK

BDH4580-4LCS 887.33 USD
BDH4580-4L
4-Methyl-2-pentanone ≥98.5% ACS, VWR Chemicals BDH®
4-Methyl-2-pentanone
Formula: (CH₃)₂CHCH₂COCH₃
MW: 100.16 g/mol
Boiling Pt: 117.4 °C (1013 hPa)
Melting Pt: –84 °C
Density: 0.801 g/cm³ (20 °C)
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00008938
CAS Number: 108-10-1
EINECS: 203-550-1
UN: 1245
ADR: 3,II
Merck Index: 13,05227
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d44947ef43cbc8103ae523ce5da380ef184ba09&vid=373b9075524004b95d32d3cf9698896714467e8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 98.5 %
Appearance Clear liquid
Titrable acid Max. 0.002 meq/g
Color (Pt-Co) Max. 15 APHA
Residue after evaporation Max. 0.005 %
Water Max. 0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR