(±)-1,2-Propanediol ≥99.5% ACS, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane

BDH4566-4L
BDH4566-4LCS 1197.33 USD
BDH4566-4L
(±)-1,2-Propanediol ≥99.5% ACS, VWR Chemicals BDH®
Propylene glycol
Formula: CH₃CH(OH)CH₂OH
MW: 76.1 g/mol
Boiling Pt: 187.6 °C (1013 hPa)
Melting Pt: –59 °C
Density: 1.04 g/cm³ (20 °C)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00064272
CAS Number: 57-55-6
EINECS: 200-338-0
Merck Index: 13,07947
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2af195b87767feb8d1baca34925153d6f97ae2d&vid=a598a9042847dc3ce52daed1a9326b592b163170&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.5 %
Color Max. 10 APHA
Titrable acid Max. 0.0005 meq/g
Residue after ignition Max. 0.005 %
Water Max. 0.2 %
Cl (Chloride) Max. 1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR