Trichloroethylene ≥99.5% ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: 1,1,2-Trichloroethene, Trichloroethene, 1,1,2-Trichloroethylene

BDH4564-4L
BDH4564-4LCS 931.95 USD
BDH4564-4L
Trichloroethylene ≥99.5% ACS, VWR Chemicals BDH®
Trichloroethylene
Formula: ClCH=CCl₂
MW: 131.39 g/mol
Boiling Pt: 86.7
Melting Pt: -73
Density: 1.46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000838
CAS Number: 79-01-6
EINECS: 201-167-4
UN: 1710
ADR: 6.1 III
Merck Index: 12,09769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ac57d7a9ea2e3097408c3559993eb33587aec09&vid=767d7d09973e747c5a2d07a4d46dffedfb559ceb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.5 %
Free halogens Passes test
Color (Pt-Co) Max. 10 APHA
Titrable acid Max. 0.0001 meq/g
Titrable base Max. 0.0003 meq/g
Heavy metals (as Pb) Max. 1 ppm
Residue after evaporation Max. 0.001 %
Water Max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR