Silver sulfate ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Disilver sulfate, Silver(I) sulfate, Disilver sulphate

BDH4560-100G
BDH4560-100GEA 838.74 USD
BDH4560-100G
Silver sulfate ACS, VWR Chemicals BDH®
Silver sulphate

Powder.

Formula: Ag₂SO₄
MW: 311.8 g/mol
Boiling Pt: 1085 °C (1013 hPa)
Melting Pt: 655 °C
Density: 5.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003407
CAS Number: 10294-26-5
EINECS: 233-653-7
Merck Index: 13,08603
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=ae7b50634b53507047974c2157e5e38c2eb90a20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 98.0 %
Insoluble matter and silver chloride Max. 0.02 %
Substances not precipitated by HCl Max. 0.03 %
NO3 (Nitrate) Max. 0.001 %
Fe (Iron) Max. 0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR