Formic acid 88,0 ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Aminic Acid, Methanoic acid

BDH4554-500ML
BDH4554-500MLCS 602.54 USD
BDH4554-500ML
Formic acid 88,0 ACS, VWR Chemicals BDH®
Formic acid
Formula: HCO₂H
MW: 46.03
Density: 1.22
Flash Pt: 48
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 88.0 %
Dilution test Passes test
SO3 (Sulphite) Passes test
Color (Pt-Co) Max. 15 APHA
Acetic acid Max. 0.4 %
Heavy metals (as Pb) Max. 5 ppm
Residue after evaporation Max. 0.002 %
Cl (Chloride) Max. 0.001 %
NH4 (Ammonium) Max. 0.005 %
SO4 (Sulphate) Max. 0.002 %
Fe (Iron) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR