Diethyl ether, anhydrous ≥99.0% stabilized, ARISTAR® ACS, VWR Chemicals BDH®

Supplier: BDH
Danger

Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane, Ethyl ether

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

203500-80405
BDH3116-1LCS 1304.4 USD
BDH3116-1L BDH3116-1L-1
Diethyl ether, anhydrous ≥99.0% stabilized, ARISTAR® ACS, VWR Chemicals BDH®
Diethyl ether
Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 35 °C (1013 hPa)
Melting Pt: –123 °C
Density: 0.7135 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Peroxide (as H₂O₂) 1 ppm max.
Preservative (BHT) Level Record only
Residue After Evaporation 0.001% max.
Titrable Acid 0.0002 meq/g max.
Water (H₂O) 0.03% max.
Assay ((CH₃CH₂)₂O) 99.0% min.
Carbonyl (as HCHO) 0.001% max.
Color (APHA) 10 max.
Alcohol (CH₃CH₂OH) To pass test (0.05%)
Identification:IR To Pass Test
Manufacture Date Record Actual
Use by Date Record Actual

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR