Sulfuric acid 95-98% ACS, FCC, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
566600-80401
566600-80601
566600-21001
566600-22001
566600-80602
566600-80603
566600-80605
BDH3074-3.8LPCS
459.37
USD
BDH3074-3.8LP
BDH3068-500MLP
BDH3076-19L
BDH3090-185L
BDH3070-2.5LPC
BDH3072-2.5LG
BDH3132-2.2LP
Sulfuric acid 95-98% ACS, FCC, VWR Chemicals BDH®
Sulfuric acid
Free from particulates.
Meets reagent specifications for testing in FCC monographs.
Formula:
H₂SO₄ MW: 98.08 g/mol Boiling Pt: ∼335 °C (1013 hPa) Melting Pt: 3 °C Density: 1.84 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 1830 ADR: 8,II Merck Index: 14,08974 |
Specification Test Results
Ammonium (NH4) | 2 ppm max. |
Appearance | Clear, colorless solution |
Arsenic (As) | 0.01 ppm max. |
Assay (H2SO4) | 95.0 - 98.0% |
Chloride (as Cl) | 0.2 ppm max. |
Color (APHA) | 10 max. |
Free from suspended matter | To pass test |
Heavy metals (as Pb) | 1 ppm max. |
Identification: sulfate per FCC | To pass test |
Iron (Fe) | 0.2 ppm max. |
Mercury (Hg) | 5 ppb max. |
Nitrate (NO3) | 0.5 ppm max. |
Residue after ignition | 5 ppm max. |
Selenium (Se) | 0.002% max. |
Substances reducing permanganate (ACS Method) | To pass test |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...