Sulfuric acid 95-98% ACS, FCC, VWR Chemicals BDH®

Supplier: VWR International
Danger

566600-80401 566600-80601 566600-21001 566600-22001 566600-80602 566600-80603 566600-80605
BDH3074-3.8LPCS 459.37 USD
BDH3074-3.8LP BDH3068-500MLP BDH3076-19L BDH3090-185L BDH3070-2.5LPC BDH3072-2.5LG BDH3132-2.2LP
Sulfuric acid 95-98% ACS, FCC, VWR Chemicals BDH®
Sulfuric acid

Free from particulates.


Meets reagent specifications for testing in FCC monographs.

Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Merck Index: 14,08974

Order Now

Specification Test Results

Ammonium (NH4) 2 ppm max.
Appearance Clear, colorless solution
Arsenic (As) 0.01 ppm max.
Assay (H2SO4) 95.0 - 98.0%
Chloride (as Cl) 0.2 ppm max.
Color (APHA) 10 max.
Free from suspended matter To pass test
Heavy metals (as Pb) 1 ppm max.
Identification: sulfate per FCC To pass test
Iron (Fe) 0.2 ppm max.
Mercury (Hg) 5 ppb max.
Nitrate (NO3) 0.5 ppm max.
Residue after ignition 5 ppm max.
Selenium (Se) 0.002% max.
Substances reducing permanganate (ACS Method) To pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR