Ammonia solution 28 - 30%, ARISTAR® ACS, VWR Chemicals BDH®

Supplier: BDH
Danger

048500-24401 BDH3014-500MLP BDH3016-2.5LG BDH3018-2.5LPC BDH3020-3.8LP BDH3126-2.2LP
BDH3022-194LEA 2339.3 USD
BDH3022-194L BDH3014-500MLP BDH3016-2.5LG BDH3018-2.5LPC BDH3020-3.8LP BDH3126-2.2LP
Ammonia solution 28 - 30%, ARISTAR® ACS, VWR Chemicals BDH®
Ammonia solution

Clear and free from particulates.


Clear and free from particulates.

Formula: NH₃
Boiling Pt: 32…37.7 °C (1013 hPa)
Melting Pt: –60…–57.5 °C
Density: 0.88…0.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless, free from susp. matter
Assay (NH3) 28.0 - 30.0%
Carbon dioxide (CO2) 0.002% max.
Chloride (Cl) 0.5 ppm max.
Heavy Metals (as Pb) 0.5 ppm max.
Iron (Fe) 0.2 ppm max.
Nitrate (NO3) 2 ppm max.
Phosphate (PO4) 2 ppm max.
Residue After Ignition 0.002% max.
Substances Reducing Permangana To pass test
Sulfate (SO4) 2 ppm max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR