Methyl ethyl ketone ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: EK 500 Additive, Butan-2-one, Ethyl methyl ketone, 2-Butanone, Butanone, MEK

BDH25643.400
BDH25643.400CS 1387.98 USD
BDH25643.400
Methyl ethyl ketone ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Methyl ethyl ketone
Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 79.6 °C (1013 hPa)
Melting Pt: –86 °C
Flash Pt: –4 °C
MDL Number: MFCD00011648
CAS Number: 78-93-3
UN: 1193
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC-FID) Min 99.5%
Color Max 10 APHA
Residue after evaporation Max 5 ppm
Trace organic impurity sec-Butyl Alcohol Max 0.2%
Titrable acid Max 0.0005 meq/g
Water (by KF coulometry) Max 0.8%
Transmittance at 329 nm Min 10 T(%)
Transmittance at 335 nm Min 30 T(%)
Transmittance at 340 nm Min 80 T(%)
Transmittance at 344 nm Min 90 T(%)
Transmittance at 350 nm Min 98 T(%)
Transmittance at 375 nm Min 98 T(%)
Transmittance at 400 nm Min 98 T(%)
Optical absorbance at 329 nm Max 1.00 AU
Optical absorbance at 335 nm Max 0.50 AU
Optical absorbance at 340 nm Max 0.10 AU
Optical absorbance at 344 nm Max 0.05 AU
Optical absorbance at 350 nm Max 0.01 AU
Optical absorbance at 375 nm Max 0.01 AU
Optical absorbance at 400 nm Max 0.01 AU

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR