N-Methyl-2-pyrrolidone (NMP), Semi Grade for the electronics industry, VWR Chemicals BDH®

Supplier: BDH
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, N-Methylpyrrolidinone, N-Methyl-2-pyrrolidinone, N-Methyl-alpha-pyrrolidone, M-Pyrol, N-methylpyrrolidone, N-Methyl-gamma-butyrolactam, 1-Methyl-2-pyrrolidone, NMP, N-Methylpyrrolidone, N-Methyl-2-pyrrolidone, Methyl Pyrrolidone

28016162
BDH2037-1GLPCS 1172.03 USD
BDH2037-1GLP
N-Methyl-2-pyrrolidone (NMP), Semi Grade for the electronics industry, VWR Chemicals BDH®
N-Methyl-2-pyrrolidone (NMP)
Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: –24 °C
Density: 1.033 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (CH3N(CH2)3CO) 99.5% Min.
Color (APHA) 50 APHA Max.
Residue after Ignition 5 ppm Max.
Water (H2O) 0.05% Max.
Chloride (Cl) 1000 ppb Max.
Phosphate (PO4) 5000 ppb Max.
Arsenic and Antimony 100 ppb Max.
Al Impurities 100 ppb Max.
B Impurities 100 ppb Max.
Cr Impurities 100 ppb Max.
Cu Impurities 100 ppb Max.
Au Impurities 100 ppb Max.
Fe Impurities 100 ppb Max.
Mg Impurities 100 ppb Max.
Mn Impurities 100 ppb Max.
Ni Impurities 100 ppb Max.
K Impurities 100 ppb Max.
Na Impurities 100 ppb Max.
Pb Impurities 100 ppb Max.
Sn Impurities 100 ppb Max.
Ti Impurities 100 ppb Max.
Zn Impurities 100 ppb Max.
Particles ≥1.0 µm 10/mL Max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR