Ethylene glycol ≥99.0%, Semi Grade for the electronics industry, VWR Chemicals BDH®

Supplier: VWR International
Warning

Synonyms: Monoethylene glycol, Ethane-1,2-diol, 2-Hydroxyethanol, Ethylene alcohol, Ethanediol, Glycol alcohol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, 1,2-Dihydroxyethane, Glycol, Dihydroxyethane

28018927
BDH2033-1GLPCS 310.4 USD
BDH2033-1GLP
Ethylene glycol ≥99.0%, Semi Grade for the electronics industry, VWR Chemicals BDH®
Ethylene glycol
Formula: HOCH₂CH₂OH
MW: 62.07 g/mol
Boiling Pt: 196…198 °C (1013 hPa)
Melting Pt: –13 °C
Density: 1.115 g/cm³ (20 °C)
Flash Pt: 116 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002885
CAS Number: 107-21-1
EINECS: 203-473-3
REACH: 01-2119456816-28
Merck Index: 13,03832
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=2002300f174929a030d66d9f1394eb6bee89b7dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (C6H6O2) 99.0% Min.
Color (APHA) 10 APHA Max.
Acidity 0.01% Max.
Water (H2O) 0.1% Max.
Boiling Range 194 ºC to 200 ºC
Specific Gravity at 25ºC 1.112 to 1.120
Appearance To Pass Test
Al Impurities 100 ppb Max.
B Impurities 100 ppb Max.
Ca Impurities 100 ppb Max.
Cr Impurities 100 ppb Max.
Cu Impurities 100 ppb Max.
Fe Impurities 100 ppb Max.
Mg Impurities 100 ppb Max.
Mn Impurities 100 ppb Max.
Ni Impurities 100 ppb Max.
K Impurities 100 ppb Max.
Na Impurities 100 ppb Max.
Pb Impurities 100 ppb Max.
Sn Impurities 100 ppb Max.
Zn Impurities 100 ppb Max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR