2-Propanol ≥70% USP, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
Synonyms:
Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol
28029711
28029720
28029741
BDH2030-1GLPCS
359.2
USD
BDH2030-1GLP
BDH2030-55GLP
BDH2030-20L
2-Propanol ≥70% USP, VWR Chemicals BDH®
2-Propanol
Filtered at 0.1 micron.
Formula:
(CH₃)₂CHOH MW: 60.1 g/mol Density: 0.8…0.92 g/cm³ (20 °C) Flash Pt: 12 Storage Temperature: Ambient |
MDL Number:
MFCD00011674 CAS Number: 67-63-0 EINECS: 200-661-7 UN: 1987 ADR: 3,II |
|
Specification Test Results
Assay (% vol) | 68.0% – 72.0% |
Specific Gravity at 20/20°C | 0.872 – 0.883 |
Description | To Pass |
Solubility | To Pass |
Identification by IR | To Pass |
Limit of Nonvolatile Residue |
|
Acidity (mL of 0.02 NaOH) |
|
Residual Solvents | To Pass |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...