Methanol ≥99.9%, Semi Grade for the electronics industry, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Wood alcohol, Methyl alcohol, Methyl hydroxide, Hydroxy methane, Carbinol, Methylol

28016347
BDH2029-1GLPCS 308.5 USD
BDH2029-1GLP
Methanol ≥99.9%, Semi Grade for the electronics industry, VWR Chemicals BDH®
Methanol
Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (CH3OH) 99.9% Min.
Color (APHA) 10 APHA Max.
Acidity 0.3 µeq/g Max.
Alkalinity 0.1 µeq/g Max.
Residue after Evaporation 5 ppm Max.
Water (H2O) 0.05% Max.
Solubility in Water To Pass Test
Chloride (Cl) 200 ppb Max.
Phosphate (PO4) 500 ppb Max.
Arsenic and Antimony 10 ppb Max.
Al Impurities 100 ppb Max.
B Impurities 100 ppb Max.
Ca Impurities 100 ppb Max.
Cr Impurities 100 ppb Max.
Cu Impurities 100 ppb Max.
Au Impurities 100 ppb Max.
Fe Impurities 100 ppb Max.
Mg Impurities 100 ppb Max.
Mn Impurities 100 ppb Max.
Ni Impurities 100 ppb Max.
K Impurities 100 ppb Max.
Na Impurities 100 ppb Max.
Pb Impurities 100 ppb Max.
Sn Impurities 100 ppb Max.
Ti Impurities 100 ppb Max.
Zn Impurities 100 ppb Max.
Particles ≥1.0 µm 10/mL Max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR