2-Propanol ≥99.5%, Semi Grade for the electronics industry, VWR Chemicals BDH®

Supplier: BDH
Danger

28015309
BDH2027-1GLPCS 266.83 USD
BDH2027-1GLP
2-Propanol ≥99.5%, Semi Grade for the electronics industry, VWR Chemicals BDH®
2-Propanol

For use in semiconductor and microelectronics manufacturing.

Formula: (CH₃)₂CHOH
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (C3H8O) 99.5% Min.
Color (APHA) 10 APHA Max.
Acidity 0.3 µeq/g Max.
Alkalinity 0.1 µeq/g Max.
Residue after Evaporation 5 ppm Max.
Water (H2O) 0.05% Max.
Chloride (Cl) 200 ppb Max.
Phosphate (PO4) 500 ppb Max.
Arsenic and Antimony 10 ppb Max.
Al Impurities 100 ppb Max.
B Impurities 10 ppb Max.
Ca Impurities 100 ppb Max.
Cr Impurities 100 ppb Max.
Cu Impurities 100 ppb Max.
Au Impurities 100 ppb Max.
Fe Impurities 100 ppb Max.
Mg Impurities 100 ppb Max.
Mn Impurities 100 ppb Max.
Ni Impurities 100 ppb Max.
K Impurities 100 ppb Max.
Na Impurities 100 ppb Max.
Pb Impurities 100 ppb Max.
Sn Impurities 100 ppb Max.
Ti Impurities 100 ppb Max.
Zn Impurities 100 ppb Max.
Particles ≥1.0 μm 10/mL Max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR