N-Methyl-2-pyrrolidone (NMP) ACS for production, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, N-Methylpyrrolidinone, N-Methyl-2-pyrrolidinone, N-Methyl-alpha-pyrrolidone, M-Pyrol, N-methylpyrrolidone, N-Methyl-gamma-butyrolactam, 1-Methyl-2-pyrrolidone, NMP, N-Methylpyrrolidone, N-Methyl-2-pyrrolidone, Methyl Pyrrolidone

BDH2022-55GLP
BDH2022-55GLPEA 10686.15 USD
BDH2022-55GLP
N-Methyl-2-pyrrolidone (NMP) ACS for production, VWR Chemicals BDH®
N-Methyl-2-pyrrolidone (NMP)

Meets ACS specifications.

Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202
Melting Pt: -24
Density: 1.033 g/cm³ (20 °C)
Flash Pt: 91
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (CH3N(CH2)3CO) 99.9% Min.
Chloride (Cl) 1000 ppb Max.
Color (APHA) 30 APHA Max.
Phosphate (PO4) 5000 ppb Max.
Water (H2O) 0.05% Max.
Free Amines 0.01% Max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR