Xylene (mixture of isomers) ACS for production, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
BDH2000-55GLM
BDH2000-5GLMS
BDH2000-55GLMEA
4028.42
USD
BDH2000-55GLM
BDH2000-5GLMS
Xylene (mixture of isomers) ACS for production, VWR Chemicals BDH®
Xylene (mixture of isomers)
Contains a maximum of 25% ethylbenzene.
- Assay percent range of greater than 98.5%
- Certified ACS reagent grade
- Available in a variety of volumes
Mixture of ortho, meta, and para isomers and may contain some Ethylbenzene.
Meets ACS specifications.
Formula:
C₆H₄(CH₃)₂ Boiling Pt: 140 °C (1013 hPa) Melting Pt: –34 °C Density: 0.86…0.88 g/cm³ (20 °C) Flash Pt: 25 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077264 CAS Number: 1330-20-7 EINECS: 215-535-7 UN: 1307 ADR: 3,III Merck Index: 13,10136 |
|
Specification Test Results
Assay by GC (Ethylbenzene) | 25.0% Max. |
GC Purity (Xylenes + Ethylbenzene) | 98.5% Min. |
Color (APHA) | 10 APHA Max. |
Residue upon Evaporation | 0.002% Max. |
Substances Darkened by Sulfuric Acid | Passes Test |
Sulfur Compounds (as S) | 0.003% Max. |
Water | 0.05% Max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...