Potassium dihydrogen phosphate ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
Synonyms:
Monopotassiumphosphate, Potassium orthophosphate monobasic, prim.-Potassium phosphate, Potassium dihydrogenorthophosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate
BDHVBDH153184U
BDH153184UEA
113.85
USD
BDH153184U
Potassium dihydrogen phosphate ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Potassium dihydrogen phosphate
Formula:
KH₂PO₄ MW: 136.09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011401 CAS Number: 7778-77-0 EINECS: 231-913-4 |
Specification Test Results
Assay | Min.99.5% |
pH (20 °C; 5 %) | 4.3-4.5 |
Heavy metals (as Pb) | Max. 10 ppm |
Cl (Chloride) | Max. 5 ppm |
SO₄ (Sulfate) | Max. 30 ppm |
Fe (Iron) | Max. 10 ppm |
Transmittance (254 nm) (0.1 mol/l) | Min. 90 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...