1-Pentanesulfonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
Synonyms:
Sodium 1-pentanesulphonate, Sodium pentane-1-sulphonate, Pentane-1-sulphonic acid sodium salt, Sodium pentanesulphonate, 98%, Sodium pentanesulphonate, Pentane-1-sulfonic acid sodium salt, Sodium pentanesulfonate, Sodium 1-pentanesulfonate, Sodium pentane-1-sulfonate, Sodium pentanesulfonate, 98%
BDHVBDH152813W
BDH152813WEA
544.93
USD
BDH152813W
1-Pentanesulfonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
1-Pentanesulphonic acid sodium salt
Ion pairing reagent.
Formula:
C₅H₁₁O₃SNa MW: 174.2 g/mol Melting Pt: > 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007541 CAS Number: 22767-49-3 EINECS: 245-208-4 |
Specification Test Results
Assay | Min. 99 % |
IR Spectrum | Passes test |
pH (10 %) | 5.5-7.5 |
Loss on drying (120°C; under vacuum) | Max. 2.0 % |
Transmittance (200 nm) (0,005 mol/l) | Min. 70 % |
Transmittance (220 nm) (0,005 mol/l) | Min. 90 % |
Transmittance (250 nm) (0,005 mol/l) | Min. 96 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...