1-Octanesulfonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International
Warning

BDH152803U
BDH152803UEA 544.18 USD
BDH152803U
1-Octanesulfonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
1-Octanesulphonic acid sodium salt

Ion pairing reagent.

Formula: C₈H₁₇NaO₃S
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 5324-84-5
EINECS: 226-195-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1244cb11db07775dec2e193823e40080024b7036&vid=32d95cd9e321818e2f94c8261c4b4e6180ee0fc7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
IR Spectrum Passes test
pH (10 %) 5.5-7.5
Loss on drying (120°C; under vacuum) Max. 2.0 %
Transmittance (200 nm) (0.005 mol/l) Min. 70 %
Transmittance (220 nm) (0.005 mol/l) Min. 90 %
Transmittance (250 nm) (0.005 mol/l) Min. 96 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR