1-Hexanesulfonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
Synonyms:
Sodium hexane-1-sulfonate, Sodium hexane-1-sulphonate, Sodium hexanesulfonate, Sodium hexanesulphonate, Sodium 1-hexanesulfonate, Sodium 1-hexanesulphonate
BDHVBDH152793M
BDH152793MEA
542.91
USD
BDH152793M
1-Hexanesulfonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
1-Hexanesulphonic acid sodium salt
Ion pairing reagent.
Formula:
C₆H₁₃O₃SNa MW: 188.22 g/mol Melting Pt: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007542 CAS Number: 2832-45-3 EINECS: 220-601-3 |
Specification Test Results
Assay | Min. 99 % |
IR Spectrum | Passes test |
pH (10 %) | 5.5-7.5 |
Loss on drying (120°C; under vacuum) | Max. 2.0 % |
Transmittance (200 nm) (0.005 mol/l) | Min. 70 % |
Transmittance (220 nm) (0.005 mol/l) | Min. 90 % |
Transmittance (250 nm) (0.005 mol/l) | Min. 96 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...