1-Heptanesulfonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International

BDH152783K
BDH152783KEA 543.94 USD
BDH152783K
1-Heptanesulfonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
1-Heptanesulphonic acid sodium salt

Ion pairing reagent.

Formula: H₃C(CH₂)₅CH₂SO₃Na
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007543
CAS Number: 22767-50-6
EINECS: 245-210-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95c09a5691140bab7446d88919f01593e8f0b9b2&vid=7c237cde5db01d0567ff19964ca3287434747480&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
IR Spectrum Passes test
pH (10 %) 5.5-7.5
Loss on drying (120°C; under vacuum) Max. 2.0 %
Transmittance (200 nm) (0.005 mol/l) Min. 70 %
Transmittance (220 nm) (0.005 mol/l) Min. 90 %
Transmittance (250 nm) (0.005 mol/l) Min. 96 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR