Ethanol denatured (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
Clear, colorless liquid. Denatured with 3.8–4.5% methanol and 4.0–5.0% 2-propanol. Suitable for cytology and histology.
- Total alcohol: 90%
- Ethyl alcohol: 81%
- Denaturant: 3.8–4.5% methanol and 4.0–5.0% 2-propanol.
Ordering information: The purchase of Reagent Alcohol in containers larger than 4 liters requires a Letterhead Permit approval by the United States Treasury Tax and Trade Bureau (TTB).
Formula:
H₃CCH₂OH Boiling Pt: 78.3 °C (1013 hPa) Flash Pt: 12 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00003568 CAS Number: 64-17-5 EINECS: 200-578-6 UN: 1170 ADR: 3,II |
Specification Test Results
Appearance | Clear, colorless liquid |
Water | 9.0 – 11.0% |
Residue | Less than 0.001% (0.0010g/124mL) |
GC Purity (Ethanol) | 81.0-82.0% |
GC Purity (Methanol) | 3.8-4.5% |
GC Purity (Isopropyl Alcohol) | 4.0-5.0% |
Color (APHA) | Less than 10 |
Specific gravity @ 20ºC | 0.822-0.829 |
Acidity | Less than 0.0025% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...