Reagent alcohol ACS (denatured with methanol and isopropanol), VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH1156-1LP BDH1156-4LP BDH1156-5GL BDH1156-19L
BDH1156-1LPCS 267.19 USD
BDH1156-1LP BDH1156-4LP BDH1156-5GL BDH1156-19L
Reagent alcohol ACS (denatured with methanol and isopropanol), VWR Chemicals BDH®
Alcohol

Meets ACS specifications for general use.


  • Total alcohol: 99.5%
  • Ethyl alcohol: 89-91%
  • Denaturant: 4-6% methyl alcohol and 4-6% 2-propanol


Ordering information: The purchase of Reagent Alcohol in containers larger than 4 liters requires a Letterhead Permit approval by the United States Treasury Tax and Trade Bureau (TTB).

Formula: H₃CCH₂OH
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear, colorless liquid
Water (K. F.) Less than 0.50%
Residue Less than 0.001% (0.0010 g/124 mL)
Assay (Ethanol + Methanol) 94.0 – 96.0 %
Assay (Isopropyl Alcohol) 4.0-6.0%
Color (APHA) Less than 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR