2,2,4-Trimethylpentane ≥99% ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane

BDHVBDH1155-19L
BDH1155-19LEA 1014.68 USD
BDH1155-19L
2,2,4-Trimethylpentane ≥99% ACS, VWR Chemicals BDH®
2,2,4-Trimethylpentane

Clear, colorless liquid. Meets ACS specifications for general use.

Formula: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Boiling Pt: 99.2 °C (1013 hPa)
Melting Pt: –107 °C
Density: 0.692 g/cm³ (20 °C)
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3,II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless liquid
Water (K.F.) Less than 0.010%
Residue Less than 0.001% (0.0010 g/145 mL)
GC Purity Greater than 99.0%
Color (APHA) Less than 10
Titrable acid Less than 0.3µeq/g (No more than 1.0mL of 0.01N sodium hydroxide)
Sulfur compounds Less than 0.005% as S

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR