2-Propanol ≥99.5% ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol

BDHVBDH1133-4LG BDHVBDH1133-19L BDHVBDH1133-1LP BDHVBDH1133-4LP PL323ZA-204 BDHVBDH1133-5GL
BDH1133-4LGCS 527.51 USD
BDH1133-4LG BDH1133-19L BDH1133-1LP BDH1133-4LP BDH1133-204L BDH1133-5GL
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
2-Propanol

Suitable for histology and cytology applications. Soluble in water. Meets ACS Specifications for general use.

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear
Water (K.F.) Less than 0.20%
Residue Less than 0.001% (0.0010 g/128 mL)
GC Purity Greater than 99.5%
Solubility in water Passes test (Solution should be as clear as an equal volume of water.)
Color (APHA) Less than 10
Titrable acid or base Less than 0.1 meq/g (No more than 0.10 mL of either 0.01N sodium hydroxide or 0.01N hydochloric acid)
Carbonyl compounds Less than 0.002% each (as acetone or propionaldehyde)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR