2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
BDH1133-4LG
BDH1133-19L
BDH1133-1LP
BDH1133-4LP
BDH1133-204L
BDH1133-5GL
BDH1133-4LGCS
527.51
USD
BDH1133-4LG
BDH1133-19L
BDH1133-1LP
BDH1133-4LP
BDH1133-204L
BDH1133-5GL
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
2-Propanol
Suitable for histology and cytology applications. Soluble in water. Meets ACS Specifications for general use.
Formula:
(CH₃)₂CHOH Boiling Pt: 82 °C (1013 hPa) Melting Pt: –89 °C Density: 0.786 g/cm³ (20 °C) Flash Pt: 12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011674 CAS Number: 67-63-0 EINECS: 200-661-7 UN: 1219 ADR: 3,II REACH: 01-2119457558-25 Merck Index: 13,05228 |
|
Specification Test Results
Appearance | Clear |
Water (K.F.) | Less than 0.20% |
Residue | Less than 0.001% (0.0010 g/128 mL) |
GC Purity | Greater than 99.5% |
Solubility in water | Passes test (Solution should be as clear as an equal volume of water.) |
Color (APHA) | Less than 10 |
Titrable acid or base | Less than 0.1 meq/g (No more than 0.10 mL of either 0.01N sodium hydroxide or 0.01N hydochloric acid) |
Carbonyl compounds | Less than 0.002% each (as acetone or propionaldehyde) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...