Diethyl ether ≥98% stabilized ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane, Ethyl ether

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

BDHVBDH1121-19L BDHVBDH1121-1LPC PL108ZA-4PC
BDH1121-19LEA 731.12 USD
BDH1121-19L BDH1121-1LPC BDH1121-4LPC
Diethyl ether ≥98% stabilized ACS, VWR Chemicals BDH®
Diethyl ether

Suitable for organic synthesis, cleaning, and prep scale synthesis. Contains ~1ppm BHT as preservative.

Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 35 °C (1013 hPa)
Melting Pt: –123 °C
Density: 0.7135 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless liquid
Water (K.F.) Less than 0.030%
Residue Less than 0.001% (0.0010 g/141 ml)
GC Purity Greater than 99.0%
Peroxides (TiCl₄; mg/L) Less than 1.0 mg/L as H₂O₂ (Typical - Not detected (less than 0.1 mg/L))
Peroxides (TiCl₄; mg/Kg) Less than 1.0 mg/Kg as H₂O₂ (Typical - Not detected (less than 0.1 mg/Kg))
Color (APHA) Less than 10
Titrable acid Less than 0.2 meq/g (Not more than 0.30 ml of 0.01N sodium hydroxide)
Carbonyl (as HCHO) Less than 0.001% (The total diffusion current should not be greater than ½ that of sample plus aldehyde)
Alcohol (as CH₂CH₃OH) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR