N,N-Dimethylformamide, low water content ≥99.5% ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Formic acid dimethylamide, DMFA, Formyldimethylamine, N-Formyldimethylamine, Dimethylformamide, NSC 5356, DMF

BDHVBDH1119-19L
BDH1119-19LEA 1775.69 USD
BDH1119-19L
N,N-Dimethylformamide, low water content ≥99.5% ACS, VWR Chemicals BDH®
N,N-Dimethylformamide

Low water. Low amine content.

Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: –61 °C
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 57.5 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless liquid (can be slightly yellow)
Water (K.F.) Less than 300 ppm
GC Purity Greater than 99.5%
Color (APHA) Less than 15
Amines [as dimethylamine (DMA)] Less than 10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR