Cyclohexane ≥99% ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH1111-4LG BDH1111-19L
BDH1111-4LGCS 949.42 USD
BDH1111-4LG BDH1111-19L
Cyclohexane ≥99% ACS, VWR Chemicals BDH®
Cyclohexane

Clear liquid.

Formula: C₆H₁₂
Boiling Pt: 81 °C (1013 hPa)
Melting Pt: 7 °C
Density: 0.7785 g/cm³ (20 °C)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear
Water (K.F.) Less than 0.02%
Residue Less than 0.002% (0.0020 g/129 mL)
GC Purity Greater than 99.0%
Color (APHA) Less than 10
Substances darkened by sulfuric acid Passes test (Neither the cyclohexane nor acid layer should be darkened.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR