Acetonitrile, low water content ≥99.5% ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH1105-4LG
BDH1105-4LGCS 1385.63 USD
BDH1105-4LG
Acetonitrile, low water content ≥99.5% ACS, VWR Chemicals BDH®
Acetonitrile

Clear liquid. Packaged in a glass bottle with a special 11/2-turn threaded neck to facilitate use on synthesizers.

Formula: H₃CCN
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –45.7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear
Water (K.F.) Less than 0.003%
Residue Less than 0.005% (0.0010g/50mL)
GC Purity Greater than 99.5%
Color (APHA) Less than 10
Titrable acid Less than 8 meq/g (No more than 8.0 mL of 0.01N sodium hydroxide)
Titrable base Less than 0.6 meq/g (No more than 4.5 mL of 0.01N hydrochloric acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR