Acetone 99,5 ACS, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
Synonyms:
2-Propanone, DMK, Propanone, Dimethyl ketone
BDH1101-1LP
BDH1101-4LG
BDH1101-204L
BDH1101-19L
BDH1101-4LP
BDH1101-1LPCS
346.06
USD
BDH1101-1LP
BDH1101-4LG
BDH1101-204L
BDH1101-19L
BDH1101-4LP
Acetone 99,5 ACS, VWR Chemicals BDH®
Acetone
Suitable for histology and cytology applications.
Formula:
CH₃COCH₃ MW: 58.08 g/mol Boiling Pt: 56.2 Melting Pt: -95.4 Density: 0.792 g/cm³ (20 °C) Flash Pt: <–20 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00008765 CAS Number: 67-64-1 EINECS: 200-662-2 UN: 1090 ADR: 3 II REACH: 01-2119471330-49 Merck Index: 13,00067 |
|
Specification Test Results
Appearance | Clear, colorless liquid |
Water (KF) | Less than 0.50% |
Residue | Less than 0.001% (0.0010 g/127 mL) |
GC Purity | Greater than 99.5% |
Solubility in water | Passes test (Solution remains clear for 30 minutes) |
Color (APHA) | Less than 10 |
Titrable acid | Less than 0.3 eq/g (No more than 1.0 mL of 0.01N sodium hydroxide) |
Titrable base | Less than 0.6 eq/g (No more than 1.0 mL of 0.01N hydrochloric acid) |
Aldehyde (as HCHO) | Less than 0.002% (Passes test - Turbidity of sample less than that of the standard) |
Isopropyl alcohol | Less than 0.05% |
Methanol | Less than 0.05% |
Substances reducing permanganate | Passes test (Pink solution after 15 min.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...