Acetone ≥99.5% ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

BDH1101-1LP BDH1101-4LG BDH1101-204L BDH1101-19L BDH1101-4LP
BDH1101-1LPCS 346.06 USD
BDH1101-1LP BDH1101-4LG BDH1101-204L BDH1101-19L BDH1101-4LP
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Acetone

Suitable for histology and cytology applications.

Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2
Melting Pt: -95.4
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless liquid
Water (KF) Less than 0.50%
Residue Less than 0.001% (0.0010 g/127 mL)
GC Purity Greater than 99.5%
Solubility in water Passes test (Solution remains clear for 30 minutes)
Color (APHA) Less than 10
Titrable acid Less than 0.3 eq/g (No more than 1.0 mL of 0.01N sodium hydroxide)
Titrable base Less than 0.6 eq/g (No more than 1.0 mL of 0.01N hydrochloric acid)
Aldehyde (as HCHO) Less than 0.002% (Passes test - Turbidity of sample less than that of the standard)
Isopropyl alcohol Less than 0.05%
Methanol Less than 0.05%
Substances reducing permanganate Passes test (Pink solution after 15 min.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR