N,N-Dimethylformamide-[D₇] (99.5% D) for NMR spectroscopy

Supplier: VWR International
Warning

Synonyms: Heptadeutero-N,N-dimethylformamide, N,N-Di[2H3]methyl[2H]formamide, N,N-Dimethylformamide-d7, DMF-D7, DMF-d7, N,N-Dimethylformamide-[²H₇], Formdimethylamide-D7, Formic acid dimethylamide-D7

BDH0914-500ML BDH0914-10ML
BDH0914-500MLEA 19368.42 USD
BDH0914-500ML BDH0914-10ML
N,N-Dimethylformamide-[D₇] (99.5% D) for NMR spectroscopy
N,N-Dimethylformamide-[D7]

Deuterium Labeled Compounds. These high-quality products are used as solvents in NMR spectroscopy, R&D, tracers, and building blocks in pharmaceutical ingredients, as well as for the synthesis of organic light-emitting diodes (OLED).


  • Highest quality deuterated stable isotopes for NMR. NMR solvents are batch tested for both chemical and isotopic purity by 1H-NMR, GC and Karl Fischer titration.

Formula: DCOON(CD₃)₂
Boiling Pt: 153 °C (1,013 hPa)
Melting Pt: –60 °C (1,013 hPa)
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00003286
CAS Number: 4472-41-7
EINECS: 224-745-8
UN: 2265
ADR: 3,III

Order Now

Specification Test Results

Appearance/color Colorless
Appearance/form Liquid
D content 1H-NMR min. 99.50 %(mol)
w(H2O) 1H-NMR max. 0.030 %(w)
1H-NMR Purity conform
H2O + D2O Content by KF Coulometer max 0.050 %(w)
GC Content min. 99.0 %(w)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR